RBC Lysis Buffer, IBI Scientific
500 ml
  89135-034
 :  IB47622
RBC Lysis Buffer, IBI Scientific
  89135-034
 :  
RBC Lysis Buffer, IBI Scientific
500 ml
 :  
$171.34
Each
RBC Lysis Buffer is used to lyse red blood cells in nucleic acid isolation applications.
  • Lyses red blood cells for DNA and RNA extractions
Can be used with the Genomic DNA (Blood) Extraction Kits and the Total RNA (Blood) Extraction Kits.
  • Taille:
    500 ml