RBC Lysis Buffer, IBI Scientific
500 ml
89135-034
: IB47622
RBC Lysis Buffer is used to lyse red blood cells in nucleic acid isolation applications.
- Lyses red blood cells for DNA and RNA extractions
Can be used with the Genomic DNA (Blood) Extraction Kits and the Total RNA (Blood) Extraction Kits.
- Taille:500 ml