RBC Lysis Buffer, IBI Scientific
135 ml
Catalog # 89135-030
Supplier: IBI Scientific
Fournisseur #: IB47620
Jump to:
Product Details & Documents
RBC Lysis Buffer is used to lyse red blood cells in nucleic acid isolation applications.
- Lyses red blood cells for DNA and RNA extractions
Can be used with the Genomic DNA (Blood) Extraction Kits and the Total RNA (Blood) Extraction Kits.
Spécifications
Spécifications
- Taille:135 ml