RBC Lysis Buffer, IBI Scientific | Avantor
RBC Lysis Buffer, IBI Scientific
135 ml
Catalog # 89135-030
Supplier:   IBI Scientific
Fournisseur #:  IB47620
RBC Lysis Buffer, IBI Scientific
Catalog # 89135-030
Supplier:  IBI Scientific
CAS Number:  
RBC Lysis Buffer, IBI Scientific
135 ml
CAS Number:  
Price
$67.42
Log in pour la disponibilité
Each
Product Details & Documents
RBC Lysis Buffer is used to lyse red blood cells in nucleic acid isolation applications.
  • Lyses red blood cells for DNA and RNA extractions
Can be used with the Genomic DNA (Blood) Extraction Kits and the Total RNA (Blood) Extraction Kits.
Spécifications
Spécifications
  • Taille:
    135 ml