Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
PYREX® Solution Bottles, Corning | Avantor

PYREX® Solution Bottles, Corning

Plain, 13.2 L (3.5 gal.)
Catalog # 89091-148
Supplier:   Corning
Fournisseur #:  1595-3X
PYREX® Solution Bottles, Corning
Catalog # 89091-148
Supplier:  Corning
PYREX® Solution Bottles, Corning
PYREX® Solution Bottles, Corning

Plain, 13.2 L (3.5 gal.)

Price
$2,270.00
Log in pour la disponibilité
Case of 4
Product Details & Documents

Designed for storage of solutions or media preparation. The 13.25 L sizes are conventional shape; the 19 and 45.5 L sizes are similar to carboys in design. Necks are tooled to increase mechanical strength and for uniform stopper fit. Accepts No. 12 rubber stopper. Type I glass.

Available plain or with graduations. Graduated bottles have black enamel graduations in double scale, graduated in 500 ml increments.

Ordering information: Stopper not included.

Spécifications
  • Capacité:
    13.2 L (3.5 gal.)
  • Désignation:
    Plain