Pipette Tips, Sorenson BioScience
Extended Length, Low Binding, 1 - 300 µl, Naturel
Catalog # 89082-376
Supplier:   Sorenson Bioscience
 :  30800T
Pipette Tips, Sorenson BioScience
Catalog # 89082-376
Pipette Tips, Sorenson BioScience
Extended Length, Low Binding, 1 - 300 µl, Naturel
Price
$63.94
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1,000 Case of 10000
Product Details & Documents
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Spécifications
  • Volume:
    1 - 300 µl
  • Tip Color:
    Naturel
  • Tip Style:
    Extended Length, Low Binding