VWR® HDPE Multipurpose Containers | Avantor
VWR® HDPE Multipurpose Containers
1.9 L (64 oz.), 16.5 dia.×11.4H cm (6¹/₂×4¹/₂")
Catalog # 89009-670
Supplier:   VWR International
VWR® HDPE Multipurpose Containers
Catalog # 89009-670
Supplier:  VWR International
VWR® HDPE Multipurpose Containers
VWR® HDPE Multipurpose Containers
1.9 L (64 oz.), 16.5 dia.×11.4H cm (6¹/₂×4¹/₂")
Price
$673.03
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 50
Product Details & Documents
Translucent containers for the storage of pathology, industrial, and environmental samples.
  • HDPE construction
  • Chemical-resistant
  • Translucent siding
Containers are made of high-quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800 mL container is equipped with a handle for portability.
Documents
Spécifications
  • Capacité:
    1.9 L (64 oz.)
  • Dimensions:
    16.5 dia.×11.4H cm (6¹/₂×4¹/₂")