Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VITLAB® Dropping Bottles, LDPE, BrandTech | Avantor

VITLAB® Dropping Bottles, LDPE, BrandTech

50 ml (1.7 oz.), 37 mm, GL18
Catalog # 83009-066
Supplier:   Brandtech
Fournisseur #:  94587
VITLAB® Dropping Bottles, LDPE, BrandTech
Catalog # 83009-066
Supplier:  Brandtech
VITLAB® Dropping Bottles, LDPE, BrandTech
VITLAB® Dropping Bottles, LDPE, BrandTech

50 ml (1.7 oz.), 37 mm, GL18

Price
$250.06
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 24
Product Details & Documents

Translucent dropping bottles come with a high-density polyethylene dropper screw cap.

  • Cut-to-size dropper with attached cap
  • LDPE
  • Five different sizes

The bottles provide excellent chemical resistance to aliphatic alcohols, aldehydes, alkalis, and acids.

Spécifications
  • Taille du bouchon:
    GL18
  • Ø:
    37 mm
  • Capacité:
    50 ml (1.7 oz.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.