Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech

VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech

160.00 ml, 50×110 mm, Oui
Catalog # 83009-023
Supplier:   Brandtech
Supplier #:  69294
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
Catalog # 83009-023
Supplier:  Brandtech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech

160.00 ml, 50×110 mm, Oui

Price
140,18 $
New Customer Promotional Price
Log in for availability
Pack of 10
Product Details & Documents

General-purpose sample vials for a variety of lab applications.

  • Polypropylene
  • LDPE push-on 'snap' caps
  • Vials are autoclavable, caps are not

Translucent polypropylene vials include low-density polyethylene push-on caps. Vials and caps are suitable for microwave use.

Specifications
  • Avec bouchon:
    Oui
  • Capacité:
    160.00 ml
  • Diameter ×H:
    50×110 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.