Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VITLAB® Forceps, POM, BrandTech | Avantor

VITLAB® Forceps, POM, BrandTech

250 mm (9⅞")
Catalog # 83009-015
Supplier:   Brandtech
Fournisseur #:  68399
VITLAB® Forceps, POM, BrandTech
Catalog # 83009-015
Supplier:  Brandtech
VITLAB® Forceps, POM, BrandTech
VITLAB® Forceps, POM, BrandTech

250 mm (9⅞")

Price
$186.46
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 5
Product Details & Documents

Polyoxymethylene (POM) forceps with rounded ends.

  • Rounded ends
  • Opaque orange-yellow

POM is suitable for autoclaving but not suitable for microwaves. Frequent autoclaving may reduce mechanical stability. Resistant to aliphatic alcohols, ether, aldehydes, aliphatic hydrocarbons, aromatic hydrocarbons, halogenated hydrocarbons, ketones, and alkalis.

Operating temperature: –40 to 130°C (–40 to 266°F)

 : Not recommended for use with ester, acids, and oxidizing agents.

Spécifications
  • Longueur:
    250 mm (9⅞")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.