VITLAB® Measuring Scoops, Polypropylene, BrandTech
1 L (33.8 oz.), 38.5 cm (15⅛")
83008-696
Supplier #: 39994
One-piece, molded measuring scoops
- Polypropylene
- Hgih quality
Features approximate volumes marked on handle.
- Capacité:1 L (33.8 oz.)
- Longueur:38.5 cm (15⅛")