VITLAB® Measuring Scoops, Polypropylene, BrandTech
100 ml (3.4 oz.), 20 cm (7⅞")
  83008-690
 :   Brandtech
 :  39694
VITLAB® Measuring Scoops, Polypropylene, BrandTech
  83008-690
 :  Brandtech
VITLAB® Measuring Scoops, Polypropylene, BrandTech
VITLAB® Measuring Scoops, Polypropylene, BrandTech
100 ml (3.4 oz.), 20 cm (7⅞")
$100.46
Pack of 12
One-piece, molded measuring scoops
  • Polypropylene
  • Hgih quality
Features approximate volumes marked on handle.
  • Capacité:
    100 ml (3.4 oz.)
  • Longueur:
    20 cm (7⅞")