Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Test Tubes, Greiner Bio-One

14 ml, Without Cap, 100 mm, 17 mm
Catalog # 82050-268
Supplier:   Greiner Bio-One
 :  187101
Test Tubes, Greiner Bio-One
Catalog # 82050-268
Supplier:  Greiner Bio-One
Test Tubes, Greiner Bio-One
Test Tubes, Greiner Bio-One

14 ml, Without Cap, 100 mm, 17 mm

Price
$248.20
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1,500
Product Details & Documents

These high-quality test tubes are available in a wide variety of materials, sizes, and volume ranges.

  • Excellent optical clarity
  • Round, flat, or conical bottom designs available
  • Polycarbonate, polypropylene, or polystyrene construction available
  • Exceptional thermal and chemical stability

Test tubes provide excellent optical clarity, and are available in round or conical bottom designs. Unless otherwise noted, tube caps are not included.

 : Test tube 82050-248 is supplied sterile.

Spécifications
  • Ø:
    17 mm
  • Capacité:
    14 ml
  • Désignation:
    Without Cap
  • Hauteur:
    100 mm
Accessories

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Screw Cap for Polypropylene Tube, White, For Model 163201
    Screw Cap for Polypropylene Tube, White, For Model 163201
    10066-772
    Log in pour la disponibilité
    Case of 9,000 - $935.15