Test Tubes without Rim
10 ml, 15×125 mm
Catalog # 82024-106
Supplier: United Scientific Supplies
Fournisseur #: TT9820-D
Jump to:
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
- Tubes do not have rims
- Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Spécifications
- Capacité:10 ml
- Diameter ×H:15×125 mm