VWR® Microscope Slide Boxes for 25 Slides | Avantor
VWR® Microscope Slide Boxes for 25 Slides
Red
Catalog # 82003-422
Supplier:   VWR International
VWR® Microscope Slide Boxes for 25 Slides
Catalog # 82003-422
Supplier:  VWR International
VWR® Microscope Slide Boxes for 25 Slides
VWR® Microscope Slide Boxes for 25 Slides
Red
Price
$57.03
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each Case of 72
Product Details & Documents
These heavy-duty, two-piece slide boxes feature cork lining and store up to 25 standard 25x75 mm (1x3") slides.
  • Heavy-duty design
  • Color-coded for easy identification
Boxes stack securely on top of each other. Ideal for storage or transport. Dimensions: 14Lx8.9Wx3.2Dcm (51/2x31/2x11/4").
Documents
Spécifications
  • Color:
    Red