Storage Microplates 384-well Polypropylene, Corning®
Product Details & Documents
These 384-well polypropylene plates and blocks are built to the industry standard footprint
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks are certified DNase- and RNase-free.
Ordering information: The 384-well plates are packaged 25 per pack, except for 46610-064 which is packaged 10 per pack. The 384-well blocks are packaged 5 per pack.
Spécifications
- Sterility:Non sterile
- Well Volume:240 µL
- Color:Clear
- Désignation:V-Bottom
- Cat. No.:80081-126
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Sealing Mat
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



