Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Direct Thermal Printer Labels, TimeMed | Avantor

Direct Thermal Printer Labels, TimeMed

Permanent, single-ply, smudgeproof, litho labels, White, 25.4×57.2 mm (1×2.25"), 1000 labels/roll, 1 roll
Catalog # 73390-030
Fournisseur #:  PSLP-1214
Direct Thermal Printer Labels, TimeMed
Catalog # 73390-030
Direct Thermal Printer Labels, TimeMed
Direct Thermal Printer Labels, TimeMed

Permanent, single-ply, smudgeproof, litho labels, White, 25.4×57.2 mm (1×2.25"), 1000 labels/roll, 1 roll

Price
41,09 $
Log in pour la disponibilité
Each
Product Details & Documents

This wide array of label types serves a variety of labeling applications in medical and research laboratories.

  • All labels are white unless otherwise noted

General-purpose labels are compatible with standard direct thermal printers. Printer-specific labels are designed for exclusive use with printers from the designated manufacturer.

Spécifications
  • Color:
    White
  • Désignation:
    Permanent, single-ply, smudgeproof, litho labels
  • Dimensions:
    25.4×57.2 mm (1×2.25")
  • Packaging:
    1000 labels/roll, 1 roll