ContactUsLinkComponent
Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
  72830-422
   Low Manufacturing Impact
Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
  72830-422
 :  3501-05
   Low Manufacturing Impact
  • Volume:
    0.1 - 10 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Cat. no.:
    72830-422
  • Sterile:
    +
  • Tip Style:
    Pure 10
  • Supplier No.:
    3501-05
Molecular BioProducts low retention pipette tips are specifically designed for eliminating sample retention within the pipette tip.
All Pure tips are racked, pre-sterilized, and certified free of RNase, DNase, DNA, and pyrogen.
The inner surface of the tip is extremely hydrophobic, reducing loss of sample from adhesion to the tip interior. Tips are crystal clear to allow easy sample viewing.
Frequently Bought Together