Safety Containers
Cease fire receptacles 55 gallon, gray, 55 gal.
Catalog # 71990-006
Supplier: Justrite
Fournisseur #: 26655G
Jump to:
Product Details & Documents
Safely contain hazardous liquids and control vapors to reduce the risk of fire, protecting both personnel and facilities.
- Safely contain hazardous liquids and control vapors to reduce the risk of fire
- Helps comply with OSHA regulations and state/local fire codes
Cease-Fire® waste receptacles provide protection against trash fires.
Helps comply with OSHA regulations and state and local fire codes.
Improve efficiency with ease-of-use features which enhance pouring and filling operations.
Spécifications
- Capacité:55 gal.
- Désignation:Cease fire receptacles 55 gallon, gray
- Certification:FM, UL
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Extinguishing Head
Informations sur le produit
- Catalog #AvailabilityPrice
- Tubing, 3/8", Stainless Steel, EDPM Seals
Informations sur le produit
- Catalog #AvailabilityPrice
- Disconnect Kit Replacement Stainless Steel
Informations sur le produit
- Catalog #AvailabilityPrice
- Disconnect Kit Replacement Polypropylene
Informations sur le produit
- Catalog #AvailabilityPrice
- Stainless Steel Disconnect Replacement For 12018Stainless Steel Disconnect Replacement For 1201889178-228Log in pour la disponibilitéEach - $1,538.56
Informations sur le produit
- Catalog #AvailabilityPrice
- Funnelspout Safety Can
Informations sur le produit
- Catalog #AvailabilityPrice
- Hose Galvanized Safety Can
Informations sur le produit
- Catalog #AvailabilityPrice
- Tubing, 3/8", Stainless Steel, VITON Seals
Informations sur le produit
- Catalog #AvailabilityPrice
- Disconnect Assembly Stainless Steel, 1/4" FittingDisconnect Assembly Stainless Steel, 1/4" Fitting14222-214Log in pour la disponibilitéEach - $1,016.71
Informations sur le produit
- Catalog #AvailabilityPrice
- Tubing, 3/8", Stainless Steel, PTFE Seals
Informations sur le produit
- Catalog #AvailabilityPrice
- Tubing, 1/4", 3/8"
Informations sur le produit
- Catalog #AvailabilityPrice
- Spill Basin Hplc Can