Trypsinizing Flasks, with Cap, Wheaton
75 ml, 33-430
Catalog # 62401-850
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 355803
Jump to:
Product Details & Documents
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
Specifications
- Taille du bouchon:33-430
- Capacité:75 ml