Trypsinizing Flasks, with Cap, Wheaton
35 ml, 24-430
62401-636
Supplier #: 355442
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
- Taille du bouchon:24-430
- Capacité:35 ml