Erlenmeyer Shake Flasks with Three Baffles, Wheaton
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
250 ml
  62401-556
Supplier #:  354239
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
  62401-556
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
250 ml
$95.55
Case of 1
Designed for use on either rotary or reciprocating shakers.
  • Autoclavable
  • Specially designed to maximize aeration for improved growth
  • Pinched neck design to reduce splashing on flasks with four baffles
Feature a marking spot on the side of the flask and a break-resistant lip, which accommodates a cotton plug or stopper. Made of wheaton 33® low-extractable borosilicate glass, which meets ASTM Type I Class A and USP Type I standards.
  • Capacité:
    250 ml