Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
7-Day Chart, 0...50 °C
61280-317
: Dickson
: C663
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, and user-selectable recording times and temperature ranges
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
: Recorders include one red pen, one blue pen, AC adapter, and 9V battery. Charts must be purchased separately.
: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
- Plage de température:0...50 °C
- Désignation:7-Day Chart