Staticide® Antistatic Solution, ACL
General-Purpose, 946 mL (1 qt.)
Catalog # 58611-305
Supplier: ACL Staticide
: 2003
Jump to:
Product Details & Documents
Easy-to-use topical antistatic solution for long-term static control
Choose from pour-top gallons or ready-to-use quart bottles with sprayer. Cleanroom formula is individually bagged to prevent contamination.
Apply to floors, walls, ceilings, fixtures, work surfaces, carpets, and more. Effective in PC-board solder fault reduction programs; used in high sensitivity electronic and EDP operations. Nontoxic and nonflammable. Completely biodegradable
: Cleanroom formula is ISO Class 5 (FED-STD-209E Class 100/M3.5) approved. Meets MIL-B-81705B and NFPA-56A electrostatic decay criteria.
Certificates
Recherche par numéro de lotSpécifications
- Désignation:General-Purpose
- Taille:946 mL (1 qt.)