Wire Shelving, Chrome, Eagle MHC®
3 lbs.
  58202-188
 :  P54-C
Wire Shelving, Chrome, Eagle MHC®
  58202-188
Wire Shelving, Chrome, Eagle MHC®
Wire Shelving, Chrome, Eagle MHC®
3 lbs.
$104.97
Each Case of 4
Wire shelving features bright nickel plating topped with a chrome-plated finish.
  • Simple two-step assembly includes a positive lock created between shelf and post without the use of any tools
  • Tapered split sleeves of high temperature-resistant ABS plastic create a positive lock that becomes stronger as the load increases
Shelf construction provides a pincer hold on mat wires that adds significant strength and distributes the entire load without stress and strain on the welds.
Chrome posts are numbered in increments of 2.5 cm (1") to ensure fast and level assembly. Leveling feet are provided to help compensate for uneven floor surfaces. Post cap is supplied.
Unique QuadTruss® design provides up to 25% more strength and a retaining ledge for increased storage stability and product retention. Open-wire design promotes higher visibility, allows light and air to pass through the shelves for reducing moisture and dust build-up, and increases the effectiveness of fire suppression systems.
 : Meets MIL-S-40144E specifications. NSF-listed for dry storage environments.
 : Manufacturer's one-year limited warranty.
  • Weight:
    3 lbs.