Hard Rubber Spatulas
203 mm (8")
Supplier #: H/R-8"
Ideal for use with materials that affect metals adversely, for mixing, weighing, cleaning, blending, and many other uses.
- Non sparking
- Wide and flexible blade
Hand-finished and hand-polished blades. Spatulas are rust-proof, washable, and sterilizable.
Documents
- Blade Length:203 mm (8")