Hard Rubber Spatulas
Hard Rubber Spatulas
203 mm (8")
  57930-069
Supplier #:  H/R-8"
Hard Rubber Spatulas
  57930-069
Hard Rubber Spatulas
Hard Rubber Spatulas
203 mm (8")
$184.71
Each Pack of 12
Ideal for use with materials that affect metals adversely, for mixing, weighing, cleaning, blending, and many other uses.
  • Non sparking
  • Wide and flexible blade
Hand-finished and hand-polished blades. Spatulas are rust-proof, washable, and sterilizable.
Documents
  • Blade Length:
    203 mm (8")