Sampling Bag Envelopes
102×178 mm (4×7")
Catalog # 56775-041
Supplier: American Mail-Well Envelope
Fournisseur #: 4060
Jump to:
Product Details & Documents
Made of extra-heavy tan kraft paper with improved metal fastener for storing mineral samples.
Spécifications
- W×L:102×178 mm (4×7")