Sampling Bag Envelopes
76.2×127 mm (3×5")
Catalog # 56775-038
Supplier: American Mail-Well Envelope
Fournisseur #: 4052
Jump to:
Product Details & Documents
Made of extra-heavy tan kraft paper with improved metal fastener for storing mineral samples.
Spécifications
- W×L:76.2×127 mm (3×5")