Sampling Bag Envelopes
89×152 mm (3.5×6")
Catalog # 56775-029
Supplier: American Mail-Well Envelope
Supplier #: 4056
Jump to:
Product Details & Documents
Made of extra-heavy tan kraft paper with improved metal fastener for storing mineral samples.
Spécifications
- W×L:89×152 mm (3.5×6")