Digestion Tubes, Labconco®
250 ml, Standard Straight Tubes
Catalog # 51001-441
Supplier:   Labconco
 :  2304025
Digestion Tubes, Labconco®
Catalog # 51001-441
Supplier:  Labconco
Digestion Tubes, Labconco®
Digestion Tubes, Labconco®
250 ml, Standard Straight Tubes
Price
2 640,00 $
Log in pour la disponibilité
Pack of 25
Product Details & Documents
Precise height and controlled wall thickness ensure exhaust-system compatibility and accurate, consistent fit.
Made of thick-wall borosilicate glass with heavy-duty retention lip and contoured bottom for optimum heat transfer and minimal thermal shock. Volumetric tubes feature volumetric marking for accurate dilutions, and built-in constriction for maximum refluxing and digestion speed.
Spécifications
  • Volume:
    250 ml
  • Désignation:
    Standard Straight Tubes