Digestion Tubes, Labconco®
250 ml, Standard Straight Tubes
51001-408
: Labconco
: 2304005
Precise height and controlled wall thickness ensure exhaust-system compatibility and accurate, consistent fit.
Made of thick-wall borosilicate glass with heavy-duty retention lip and contoured bottom for optimum heat transfer and minimal thermal shock. Volumetric tubes feature volumetric marking for accurate dilutions, and built-in constriction for maximum refluxing and digestion speed.
- Volume:250 ml
- Désignation:Standard Straight Tubes