Digestion Tubes, Labconco® | Avantor
Digestion Tubes, Labconco®
250 ml, Volumetric Tubes
Catalog # 51001-248
Supplier:   Labconco
Fournisseur #:  2303025
Digestion Tubes, Labconco®
Catalog # 51001-248
Supplier:  Labconco
Digestion Tubes, Labconco®
Digestion Tubes, Labconco®
250 ml, Volumetric Tubes
Price
$2,850.00
Log in pour la disponibilité
Pack of 25
Product Details & Documents
Precise height and controlled wall thickness ensure exhaust-system compatibility and accurate, consistent fit.
Made of thick-wall borosilicate glass with heavy-duty retention lip and contoured bottom for optimum heat transfer and minimal thermal shock. Volumetric tubes feature volumetric marking for accurate dilutions, and built-in constriction for maximum refluxing and digestion speed.
Spécifications
  • Volume:
    250 ml
  • Désignation:
    Volumetric Tubes