VWR® pH Electrode Arm | Avantor
VWR® pH Electrode Arm
VWR® pH Electrode Arm
Catalog # 34105-859
Supplier:   VWR International
VWR® pH Electrode Arm
Catalog # 34105-859
Supplier:  VWR International
VWR® pH Electrode Arm
VWR® pH Electrode Arm
VWR® pH Electrode Arm
Price
$475.12
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Electrode arm moves in almost any direction, holding up to three electrodes safely and securely in selected positions.
  • Ideal for multiple pH readings
  • Accepts all standard electrodes
  • Pivots 360°, Extends 53 cm (207/8")
The arm articulates at the three points to maintain electrodes in vertical position.
Designed to accept any standard glass, reference, combination, or other electrode — even a temperature compensation probe. Weighted base and spring counter-balance permit smooth, fluid movement with superior stability.
Spécifications
  • Désignation:
    VWR® pH Electrode Arm