VWR® pH Electrode Arm
VWR® pH Electrode Arm
Catalog # 34105-859
Supplier: VWR International
Jump to:
Product Details & Documents
Electrode arm moves in almost any direction, holding up to three electrodes safely and securely in selected positions.
- Ideal for multiple pH readings
- Accepts all standard electrodes
- Pivots 360°, Extends 53 cm (207/8")
The arm articulates at the three points to maintain electrodes in vertical position.
Designed to accept any standard glass, reference, combination, or other electrode — even a temperature compensation probe. Weighted base and spring counter-balance permit smooth, fluid movement with superior stability.
Spécifications
- Désignation:VWR® pH Electrode Arm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.