Polysoft Face Shields, OP-D-OP | Avantor
Polysoft Face Shields, OP-D-OP
Polysoft Face Shield, Magenta
Catalog # 33000-452
Supplier:   OP-D-OP
Fournisseur #:  206PVMG
Polysoft Face Shields, OP-D-OP
Catalog # 33000-452
Supplier:  OP-D-OP
Polysoft Face Shields, OP-D-OP
Polysoft Face Shield, Magenta
Price
$198.92
Log in pour la disponibilité
Each
Product Details & Documents
Face shield is designed for facial protection in medical facilities and laboratories.
  • Unbreakable polycarbonate shield can be replaced
  • Cinch cord can be adjusted for a secure fit
  • Crystal clear optics
  • Polysoft band for comfort
Fits over glasses and replaces goggles and masks.
Spécifications
  • Color:
    Magenta
  • Désignation:
    Polysoft Face Shield