Polysoft Face Shields, OP-D-OP
Polysoft Face Shield, Magenta
Catalog # 33000-452
Supplier: OP-D-OP
Fournisseur #: 206PVMG
Jump to:
Product Details & Documents
Face shield is designed for facial protection in medical facilities and laboratories.
- Unbreakable polycarbonate shield can be replaced
- Cinch cord can be adjusted for a secure fit
- Crystal clear optics
- Polysoft band for comfort
Fits over glasses and replaces goggles and masks.
Spécifications
- Color:Magenta
- Désignation:Polysoft Face Shield