PYREX® Thermometer Adapters, Corning
14/20, 60 mm
  32645-312
Product available on GSA Advantage®
   Low Manufacturing Impact
 :   Corning
 :  8821-14
PYREX® Thermometer Adapters, Corning
  32645-312
 :  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Thermometer Adapters, Corning
PYREX® Thermometer Adapters, Corning
14/20, 60 mm
$49.90
Case of 1
Thermometer adapters are designed with a [ST] ground joint at one end; the other end is tooled to accommodate a thermometer.
  • Supplied with a rubber thermometer holder
Note: These adapter tubes also function as a replacement part for all Corning organic chemistry kits.
  • ST Joint No.:
    14/20
  • Longueur:
    60 mm
  • Cat. No.:
    32645-312
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact