PYREX® Thermometer Adapters, Corning
14/20, 60 mm
: Corning
: 8821-14
PYREX® Thermometer Adapters, Corning
32645-312
: Corning
Product available on GSA Advantage®
Thermometer adapters are designed with a [ST] ground joint at one end; the other end is tooled to accommodate a thermometer.
- Supplied with a rubber thermometer holder
Note: These adapter tubes also function as a replacement part for all Corning organic chemistry kits.
- ST Joint No.:14/20
- Longueur:60 mm
- Cat. No.:32645-312
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact