PYREX® Thermometer Adapters, Corning
24/40, 80 mm
Catalog # 32645-185
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
 :  8821-24
PYREX® Thermometer Adapters, Corning
Catalog # 32645-185
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Thermometer Adapters, Corning
PYREX® Thermometer Adapters, Corning
24/40, 80 mm
Price
$101.00
Log in for availability
Case of 1
Product Details & Documents
Thermometer adapters are designed with a [ST] ground joint at one end; the other end is tooled to accommodate a thermometer.
  • Supplied with a rubber thermometer holder
Note: These adapter tubes also function as a replacement part for all Corning organic chemistry kits.
Specifications
  • ST Joint No.:
    24/40
  • Longueur:
    80 mm
  • Cat. No.:
    32645-185
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact