PYREX® Thermometer Adapters, Corning
24/40, 80 mm
Supplier: Corning
: 8821-24
PYREX® Thermometer Adapters, Corning
Catalog # 32645-185
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Thermometer adapters are designed with a [ST] ground joint at one end; the other end is tooled to accommodate a thermometer.
- Supplied with a rubber thermometer holder
Note: These adapter tubes also function as a replacement part for all Corning organic chemistry kits.
Specifications
- ST Joint No.:24/40
- Longueur:80 mm
- Cat. No.:32645-185
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact