Transwell® Clear Inserts, Polyester (PET) Membrane
12, 1.12 cm², 12 mm, Polyester, 3.0 µm, 12/Plate
Supplier: Corning
: 3462
Transwell® Clear Inserts, Polyester (PET) Membrane
Catalog # 29442-080
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
- w Produce a cell culture environment that closely resembles an in vivo state
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
- Treated for optimal cell attachment
- Excellent visibility under phase contrast microscopy
- Supplied in multiple well plates
- Sterile
Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.
Specifications
- Nb de puits:12
- Taille de pore:3.0 µm
- Packaging:12/Plate
- Culture Area:1.12 cm²
- Membrane Diameter:12 mm
- Matériau membranaire:Polyester
- Cat. No.:29442-080
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



