Inlet Filter, Labconco®
Inlet Filter
Catalog # 26671-506
Supplier:   Labconco
 :  1472200
Inlet Filter, Labconco®
Catalog # 26671-506
Supplier:  Labconco
Inlet Filter, Labconco®
Inlet Filter, Labconco®
Inlet Filter
Price
$1,280.00
Log in for availability
Each
Product Details & Documents
This disposable filter prevents oil backstreaming from the vacuum pump and protects the pump from damage.
Filter life is approximately 1000 hours at 10 µm vacuum.
 : Includes filter assembly and one filter cartridge.
Specifications
  • Désignation:
    Inlet Filter