Inlet Filter, Labconco®
Inlet Filter
Catalog # 26671-506
Supplier: Labconco
: 1472200
Jump to:
Product Details & Documents
This disposable filter prevents oil backstreaming from the vacuum pump and protects the pump from damage.
Filter life is approximately 1000 hours at 10 µm vacuum.
: Includes filter assembly and one filter cartridge.
Specifications
- Désignation:Inlet Filter