WHEATON® Filter Flasks, DWK Life Sciences
Filter flask, 1000 ml, graduated, No. 8 stopper joint
: 635232
These erlenmeyer-style flasks feature heavy-wall glass to provide the mechanical strength needed for vacuum filtration.
- Side arms are #2 hose connections that accept standard 1/4” (6mm) I.D. flexible tubing for connection to a vacuum source
- Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
Connect a second flask between the filtering flask and the vacuum source to prevent entry of the filtrate into the vacuum source.
- Désignation:Filter flask, 1000 ml, graduated, No. 8 stopper joint