Conductivity Solutions, YSI
1 quart, 100,000 µmhos/cm, 0.25% at 25 °C
Catalog # 23202-343
Supplier:   YSI
 :  065274
Conductivity Solutions, YSI
Catalog # 23202-343
Supplier:  YSI
CAS Number:  
Conductivity Solutions, YSI
Conductivity Solutions, YSI
1 quart, 100,000 µmhos/cm, 0.25% at 25 °C
CAS Number:  
Price
$161.29
Log in pour la disponibilité
Each (1qt US)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Use YSI Conductivity Solutions to calibrate conductivity cell and meter together as a system for maximum measurement accuracy.
Solutions are standardized against NIST Standard Reference Material potassium chloride (KCl). Each bottle includes calibration instruments and temperature correction information. Packaged in glass quart and plastic pint bottles.
Spécifications
Spécifications
  • Taille:
    1 quart
  • Accuracy:
    0.25% at 25 °C
  • Conductivity:
    100,000 µmhos/cm