Conductivity Solutions, YSI
Use YSI Conductivity Solutions to calibrate conductivity cell and meter together as a system for maximum measurement accuracy.
Solutions are standardized against NIST Standard Reference Material potassium chloride (KCl). Each bottle includes calibration instruments and temperature correction information. Packaged in glass quart and plastic pint bottles.
- Taille:1 quart
- Accuracy:0.25% at 25 °C
- Conductivity:10,000 µmhos/cm