Tool Mounting Bar, Perfex
Individual Tool Mount, Black
Catalog # 22940-074
Supplier: Perfex
Supplier #: 4001
Jump to:
Product Details & Documents
Holds three tools in an 18" friction-fit channel, allowing instant repositioning along the bar length.
- Individual mount is also available
Securely holds a wide range of tools, up to 11/2" diameter. Mounting holes are 16" on center for convenient attachment to studs, drywall or other wall surfaces.
Specifications
- Color:Black
- Désignation:Individual Tool Mount