Tool Mounting Bar, Perfex
Tool Mounting Bar, White
Catalog # 22940-072
Supplier: Perfex
Fournisseur #: 4000
Jump to:
Product Details & Documents
Holds three tools in an 18" friction-fit channel, allowing instant repositioning along the bar length.
- Individual mount is also available
Securely holds a wide range of tools, up to 11/2" diameter. Mounting holes are 16" on center for convenient attachment to studs, drywall or other wall surfaces.
Spécifications
- Color:White
- Désignation:Tool Mounting Bar