Corning® Spinner Flasks with Vertical Sidearms, Corning | Avantor
Corning® Spinner Flasks with Vertical Sidearms, Corning
36 L, 120 mm, 45 mm
  22877-138
 :   Corning
Fournisseur #:  4512-36L
Corning® Spinner Flasks with Vertical Sidearms, Corning
  22877-138
 :  Corning
Corning® Spinner Flasks with Vertical Sidearms, Corning
36 L, 120 mm, 45 mm
$4,900.00
Case of 1
Borosilicate glass flasks are baffled to enhance agitation of contents.
  • Side baffles enhance aeration and agitation of flask contents
  • Proprietary 'stretch' impeller design ensures optimal stirring
  • Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.
Informations sur les accessoires: Proculture® spinner direct top drive motor with high torque, low rpm specifically designed to maintain constant low speed for use on top of Proculture® spinner flasks. The gearhead stirrer motor delivers 14.5 in-lbs of torque and has a maximum speed of 350 rpm.
100 mm Center Cap Assembly for Proculture® Spinner Flasks is designed for use with direct overhead drive motor, these cap assemblies required optional 8L, 15 L or 36 L paddle assemblies.
  • Capacité:
    36 L
  • Sidearm Neck O.D.:
    45 mm
  • Center Neck O.D.:
    120 mm
Accessories
  •  
    Description
    Fits Flask
  • 22877-060
    ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet
    8 L or 15 L
    Case of 1 - $583.00
  •  
    Description
    Fits Flask
  • 22877-146
    ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets
    36 L
    Case of 1 - $720.00
  •  
    Description
  • 22877-044
    ProCulture™ Fittings For Gas Delivery, with 0.3 cm (1/8") Inlet
    Case of 1 - $443.00
  •  
    Description
  • 22877-048
    ProCulture™ Fittings For Gas Delivery and Venting, with 0.3 cm (1/8") and 0.6 cm
    Case of 1 - $744.00
  •  
    Description
  • 22877-050
    Filtered Gas Venting Cap, 45 mm
    Case of 1 - $276.00
  •  
    Description
  • 22877-046
    ProCulture™ Fittings For Gas Delivery, with 0.6 cm (1/4") Inlet
    Case of 1 - $466.00
  •  
    Description
  • 89089-962
    ProCulture™ Fittings For Oxygen, Temperature, or pH Probe Insertion, 18 mm (11/16")
    Case of 1 - $377.00
  •  
    Description
    Fits Flask
  • 22877-114
    Paddle Assemblies with Direct Drive Overhead Motor
    36 L
    Case of 1 - $463.00
  •  
    Description
    Fits Flask
  • 89090-016
    ProCulture™ Fittings For Media Handling, with Two 0.6 (1/4") Inlets and High Temperature O-Ring
    8 L or 15 L
    Case of 1 - $690.00
  •  
    Description
    Fits
  • 89090-230
    Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® flasks
    Case of 1 - $4,070.00
  •  
    Description
    Fits
  • 89090-232
    Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® flasks
    Case of 1 - $5,140.00
  •  
    Description
  • 89089-676
    Direct overhead drive 100 mm center cap assembly
    Case of 1 - $2,980.00
  •  
    Description
    Fits Flask
  • 22877-058
    ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets
    8 L or 15 L
    Case of 1 - $713.00
  •  
    Description
    Fits Flask
  • 22877-120
    ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets
    36 L
    Case of 1 - $735.00
  •  
    Description
    Fits Flask
  • 22877-112
    Paddle Assemblies with Direct Drive Overhead Motor
    15 L
    Case of 1 - $465.00
  •  
    Description
  • 22877-062
    ProCulture™ Fittings For Temperature Probe Insertion, 12 mm (1/2")
    Case of 1 - $296.00
  •  
    Description
    Fits Flask
  • 22877-064
    Center Neck Cap Compression Fitting Assembly for Flasks with 0.3 cm (1/8") Tubing
    8 L and 15 L
    Case of 1 - $782.00
  •  
    Description
    Fits Flask
  • 22877-118
    ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets
    36 L
    Case of 1 - $706.00
  •  
    Description
    Fits Flask
  • 22877-110
    Paddle Assemblies with Direct Drive Overhead Motor
    8 L
    Case of 1 - $460.00
  •  
    Description
  • 89090-014
    Filtered Gas Venting Cap with Sanitary Fitting, 50 mm
    Case of 1 - $381.00
  •  
    Description
    Fits Flask
  • 22877-068
    Center Neck Cap Compression Fitting Assembly for Flasks with 0.6 cm (1/4") Tubing
    8 L and 15 L
    Case of 1 - $775.00
  •  
    Description
    Fits Flask
  • 22877-054
    ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets
    8 L or 15 L
    Case of 1 - $699.00
  •  
    Description
    Fits Flask
  • 22877-056
    ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets
    8 L or 15 L
    Case of 1 - $735.00
  •  
    Description
  • 22877-144
    ProCulture™ Fittings For pH Probe Insertion, 12 mm (1/2")
    Case of 1 - $296.00
  •  
    Description
    Fits Flask
  • 22877-148
    ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet
    36 L
    Case of 1 - $628.00
  •  
    Description
  • 22877-052
    ProCulture™ Fittings For Oxygen Probe Insertion, 12 mm (1/2")
    Case of 1 - $296.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.