Corning® Spinner Flasks with Vertical Sidearms, Corning | Avantor
Corning® Spinner Flasks with Vertical Sidearms, Corning
36 L, 100 mm, 45 mm
Catalog # 22877-132
Supplier:   Corning
Fournisseur #:  4510-36L
Corning® Spinner Flasks with Vertical Sidearms, Corning
Catalog # 22877-132
Supplier:  Corning
Corning® Spinner Flasks with Vertical Sidearms, Corning
36 L, 100 mm, 45 mm
Price
$4,680.00
Log in pour la disponibilité
Case of 1
Product Details & Documents
Borosilicate glass flasks are baffled to enhance agitation of contents.
  • Side baffles enhance aeration and agitation of flask contents
  • Proprietary 'stretch' impeller design ensures optimal stirring
  • Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.
Informations sur les accessoires: Proculture® spinner direct top drive motor with high torque, low rpm specifically designed to maintain constant low speed for use on top of Proculture® spinner flasks. The gearhead stirrer motor delivers 14.5 in-lbs of torque and has a maximum speed of 350 rpm.
100 mm Center Cap Assembly for Proculture® Spinner Flasks is designed for use with direct overhead drive motor, these cap assemblies required optional 8L, 15 L or 36 L paddle assemblies.
Spécifications
  • Capacité:
    36 L
  • Sidearm Neck O.D.:
    45 mm
  • Center Neck O.D.:
    100 mm
Accessories
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet
    ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet
    22877-060
    8 L or 15 L
    Log in pour la disponibilité
    Case of 1 - $583.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets
    ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets
    22877-146
    36 L
    Log in pour la disponibilité
    Case of 1 - $720.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ProCulture™ Fittings For Gas Delivery, with 0.3 cm (1/8") Inlet
    ProCulture™ Fittings For Gas Delivery, with 0.3 cm (1/8") Inlet
    22877-044
    Log in pour la disponibilité
    Case of 1 - $443.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ProCulture™ Fittings For Gas Delivery and Venting, with 0.3 cm (1/8") and 0.6 cm
    ProCulture™ Fittings For Gas Delivery and Venting, with 0.3 cm (1/8") and 0.6 cm
    22877-048
    Log in pour la disponibilité
    Case of 1 - $744.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Filtered Gas Venting Cap, 45 mm
    Filtered Gas Venting Cap, 45 mm
    22877-050
    Log in pour la disponibilité
    Case of 1 - $276.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ProCulture™ Fittings For Gas Delivery, with 0.6 cm (1/4") Inlet
    ProCulture™ Fittings For Gas Delivery, with 0.6 cm (1/4") Inlet
    22877-046
    Log in pour la disponibilité
    Case of 1 - $466.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ProCulture™ Fittings For Oxygen, Temperature, or pH Probe Insertion, 18 mm (11/16")
    ProCulture™ Fittings For Oxygen, Temperature, or pH Probe Insertion, 18 mm (11/16")
    89089-962
    Log in pour la disponibilité
    Case of 1 - $377.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Paddle Assemblies with Direct Drive Overhead Motor
    Paddle Assemblies with Direct Drive Overhead Motor
    22877-114
    36 L
    Log in pour la disponibilité
    Case of 1 - $463.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with Two 0.6 (1/4") Inlets and High Temperature O-Ring
    ProCulture™ Fittings For Media Handling, with Two 0.6 (1/4") Inlets and High Temperature O-Ring
    89090-016
    8 L or 15 L
    Log in pour la disponibilité
    Case of 1 - $690.00
Informations sur le produit
  • Catalog #
    Fits
    Availability
    Price
  • Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)
    89090-230
    Corning® Proculture® flasks
    Log in pour la disponibilité
    Case of 1 - $4,070.00
Informations sur le produit
  • Catalog #
    Fits
    Availability
    Price
  • Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)
    89090-232
    Corning® Proculture® flasks
    Log in pour la disponibilité
    Case of 1 - $5,140.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Direct overhead drive 100 mm center cap assembly
    Direct overhead drive 100 mm center cap assembly
    89089-676
    Log in pour la disponibilité
    Case of 1 - $2,980.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets
    ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets
    22877-058
    8 L or 15 L
    Log in pour la disponibilité
    Case of 1 - $713.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets
    ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets
    22877-120
    36 L
    Log in pour la disponibilité
    Case of 1 - $735.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Paddle Assemblies with Direct Drive Overhead Motor
    Paddle Assemblies with Direct Drive Overhead Motor
    22877-112
    15 L
    Log in pour la disponibilité
    Case of 1 - $465.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ProCulture™ Fittings For Temperature Probe Insertion, 12 mm (1/2")
    ProCulture™ Fittings For Temperature Probe Insertion, 12 mm (1/2")
    22877-062
    Log in pour la disponibilité
    Case of 1 - $296.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Center Neck Cap Compression Fitting Assembly for Flasks with 0.3 cm (1/8") Tubing
    Center Neck Cap Compression Fitting Assembly for Flasks with 0.3 cm (1/8") Tubing
    22877-064
    8 L and 15 L
    Log in pour la disponibilité
    Case of 1 - $782.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets
    ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets
    22877-118
    36 L
    Log in pour la disponibilité
    Case of 1 - $706.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Paddle Assemblies with Direct Drive Overhead Motor
    Paddle Assemblies with Direct Drive Overhead Motor
    22877-110
    8 L
    Log in pour la disponibilité
    Case of 1 - $460.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Filtered Gas Venting Cap with Sanitary Fitting, 50 mm
    Filtered Gas Venting Cap with Sanitary Fitting, 50 mm
    89090-014
    Log in pour la disponibilité
    Case of 1 - $381.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Center Neck Cap Compression Fitting Assembly for Flasks with 0.6 cm (1/4") Tubing
    Center Neck Cap Compression Fitting Assembly for Flasks with 0.6 cm (1/4") Tubing
    22877-068
    8 L and 15 L
    Log in pour la disponibilité
    Case of 1 - $775.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets
    ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets
    22877-054
    8 L or 15 L
    Log in pour la disponibilité
    Case of 1 - $699.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets
    ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets
    22877-056
    8 L or 15 L
    Log in pour la disponibilité
    Case of 1 - $735.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ProCulture™ Fittings For pH Probe Insertion, 12 mm (1/2")
    ProCulture™ Fittings For pH Probe Insertion, 12 mm (1/2")
    22877-144
    Log in pour la disponibilité
    Case of 1 - $296.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet
    ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet
    22877-148
    36 L
    Log in pour la disponibilité
    Case of 1 - $628.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ProCulture™ Fittings For Oxygen Probe Insertion, 12 mm (1/2")
    ProCulture™ Fittings For Oxygen Probe Insertion, 12 mm (1/2")
    22877-052
    Log in pour la disponibilité
    Case of 1 - $296.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.