Corning® Spinner Flasks with Vertical Sidearms, Corning
Product Details & Documents
Borosilicate glass flasks are baffled to enhance agitation of contents.
- Side baffles enhance aeration and agitation of flask contents
- Proprietary 'stretch' impeller design ensures optimal stirring
- Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.
Informations sur les accessoires: Proculture® spinner direct top drive motor with high torque, low rpm specifically designed to maintain constant low speed for use on top of Proculture® spinner flasks. The gearhead stirrer motor delivers 14.5 in-lbs of torque and has a maximum speed of 350 rpm.
100 mm Center Cap Assembly for Proculture® Spinner Flasks is designed for use with direct overhead drive motor, these cap assemblies required optional 8L, 15 L or 36 L paddle assemblies.
Spécifications
- Capacité:36 L
- Sidearm Neck O.D.:45 mm
- Center Neck O.D.:100 mm
Accessories
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") InletProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet22877-0608 L or 15 LLog in pour la disponibilitéCase of 1 - $583.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") InletsProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets22877-14636 LLog in pour la disponibilitéCase of 1 - $720.00
Informations sur le produit
- Catalog #AvailabilityPrice
- ProCulture™ Fittings For Gas Delivery, with 0.3 cm (1/8") InletProCulture™ Fittings For Gas Delivery, with 0.3 cm (1/8") Inlet22877-044Log in pour la disponibilitéCase of 1 - $443.00
Informations sur le produit
- Catalog #AvailabilityPrice
- ProCulture™ Fittings For Gas Delivery and Venting, with 0.3 cm (1/8") and 0.6 cmProCulture™ Fittings For Gas Delivery and Venting, with 0.3 cm (1/8") and 0.6 cm22877-048Log in pour la disponibilitéCase of 1 - $744.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Filtered Gas Venting Cap, 45 mm
Informations sur le produit
- Catalog #AvailabilityPrice
- ProCulture™ Fittings For Gas Delivery, with 0.6 cm (1/4") InletProCulture™ Fittings For Gas Delivery, with 0.6 cm (1/4") Inlet22877-046Log in pour la disponibilitéCase of 1 - $466.00
Informations sur le produit
- Catalog #AvailabilityPrice
- ProCulture™ Fittings For Oxygen, Temperature, or pH Probe Insertion, 18 mm (11/16")ProCulture™ Fittings For Oxygen, Temperature, or pH Probe Insertion, 18 mm (11/16")89089-962Log in pour la disponibilitéCase of 1 - $377.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Paddle Assemblies with Direct Drive Overhead MotorPaddle Assemblies with Direct Drive Overhead Motor22877-11436 LLog in pour la disponibilitéCase of 1 - $463.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with Two 0.6 (1/4") Inlets and High Temperature O-RingProCulture™ Fittings For Media Handling, with Two 0.6 (1/4") Inlets and High Temperature O-Ring89090-0168 L or 15 LLog in pour la disponibilitéCase of 1 - $690.00
Informations sur le produit
- Catalog #FitsAvailabilityPrice
- Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)89090-230Corning® Proculture® flasksLog in pour la disponibilitéCase of 1 - $4,070.00
Informations sur le produit
- Catalog #FitsAvailabilityPrice
- Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)89090-232Corning® Proculture® flasksLog in pour la disponibilitéCase of 1 - $5,140.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Direct overhead drive 100 mm center cap assemblyDirect overhead drive 100 mm center cap assembly89089-676Log in pour la disponibilitéCase of 1 - $2,980.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") InletsProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets22877-0588 L or 15 LLog in pour la disponibilitéCase of 1 - $713.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") InletsProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets22877-12036 LLog in pour la disponibilitéCase of 1 - $735.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Paddle Assemblies with Direct Drive Overhead MotorPaddle Assemblies with Direct Drive Overhead Motor22877-11215 LLog in pour la disponibilitéCase of 1 - $465.00
Informations sur le produit
- Catalog #AvailabilityPrice
- ProCulture™ Fittings For Temperature Probe Insertion, 12 mm (1/2")ProCulture™ Fittings For Temperature Probe Insertion, 12 mm (1/2")22877-062Log in pour la disponibilitéCase of 1 - $296.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Center Neck Cap Compression Fitting Assembly for Flasks with 0.3 cm (1/8") TubingCenter Neck Cap Compression Fitting Assembly for Flasks with 0.3 cm (1/8") Tubing22877-0648 L and 15 LLog in pour la disponibilitéCase of 1 - $782.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") InletsProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets22877-11836 LLog in pour la disponibilitéCase of 1 - $706.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Paddle Assemblies with Direct Drive Overhead MotorPaddle Assemblies with Direct Drive Overhead Motor22877-1108 LLog in pour la disponibilitéCase of 1 - $460.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Filtered Gas Venting Cap with Sanitary Fitting, 50 mmFiltered Gas Venting Cap with Sanitary Fitting, 50 mm89090-014Log in pour la disponibilitéCase of 1 - $381.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Center Neck Cap Compression Fitting Assembly for Flasks with 0.6 cm (1/4") TubingCenter Neck Cap Compression Fitting Assembly for Flasks with 0.6 cm (1/4") Tubing22877-0688 L and 15 LLog in pour la disponibilitéCase of 1 - $775.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") InletsProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets22877-0548 L or 15 LLog in pour la disponibilitéCase of 1 - $699.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") InletsProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets22877-0568 L or 15 LLog in pour la disponibilitéCase of 1 - $735.00
Informations sur le produit
- Catalog #AvailabilityPrice
- ProCulture™ Fittings For pH Probe Insertion, 12 mm (1/2")ProCulture™ Fittings For pH Probe Insertion, 12 mm (1/2")22877-144Log in pour la disponibilitéCase of 1 - $296.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") InletProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet22877-14836 LLog in pour la disponibilitéCase of 1 - $628.00
Informations sur le produit
- Catalog #AvailabilityPrice
- ProCulture™ Fittings For Oxygen Probe Insertion, 12 mm (1/2")ProCulture™ Fittings For Oxygen Probe Insertion, 12 mm (1/2")22877-052Log in pour la disponibilitéCase of 1 - $296.00