Corning® Spinner Flasks with Vertical Sidearms, Corning
Borosilicate glass flasks are baffled to enhance agitation of contents.
- Side baffles enhance aeration and agitation of flask contents
- Proprietary 'stretch' impeller design ensures optimal stirring
- Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.
: Proculture® spinner direct top drive motor with high torque, low rpm specifically designed to maintain constant low speed for use on top of Proculture® spinner flasks. The gearhead stirrer motor delivers 14.5 in-lbs of torque and has a maximum speed of 350 rpm.
100 mm Center Cap Assembly for Proculture® Spinner Flasks is designed for use with direct overhead drive motor, these cap assemblies required optional 8L, 15 L or 36 L paddle assemblies.
- Capacité:15 L
- Sidearm Neck O.D.:45 mm
- Center Neck O.D.:100 mm
Accessories
- DescriptionFits Flask
- 22877-060ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet8 L or 15 LCase of 1 - 583,00 $
- DescriptionFits Flask
- 22877-146ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets36 LCase of 1 - 720,00 $
- Description
- Description
- 22877-048ProCulture™ Fittings For Gas Delivery and Venting, with 0.3 cm (1/8") and 0.6 cmCase of 1 - 744,00 $
- Description
- Description
- Description
- 89089-962ProCulture™ Fittings For Oxygen, Temperature, or pH Probe Insertion, 18 mm (11/16")Case of 1 - 377,00 $
- DescriptionFits Flask
- DescriptionFits Flask
- 89090-016ProCulture™ Fittings For Media Handling, with Two 0.6 (1/4") Inlets and High Temperature O-Ring8 L or 15 LCase of 1 - 690,00 $
- DescriptionFits
- 89090-230Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® flasksCase of 1 - 4 070,00 $
- DescriptionFits
- 89090-232Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® flasksCase of 1 - 5 140,00 $
- Description
- DescriptionFits Flask
- 22877-058ProCulture™ Fittings For Media Handling, with 0.3 cm (1/8") and 0.6 cm (1/4") Inlets8 L or 15 LCase of 1 - 713,00 $
- DescriptionFits Flask
- 22877-120ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets36 LCase of 1 - 735,00 $
- DescriptionFits Flask
- Description
- DescriptionFits Flask
- 22877-064Center Neck Cap Compression Fitting Assembly for Flasks with 0.3 cm (1/8") Tubing8 L and 15 LCase of 1 - 782,00 $
- DescriptionFits Flask
- 22877-118ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets36 LCase of 1 - 706,00 $
- DescriptionFits Flask
- Description
- DescriptionFits Flask
- 22877-068Center Neck Cap Compression Fitting Assembly for Flasks with 0.6 cm (1/4") Tubing8 L and 15 LCase of 1 - 775,00 $
- DescriptionFits Flask
- 22877-054ProCulture™ Fittings For Media Handling, with Two 0.3 cm (1/8") Inlets8 L or 15 LCase of 1 - 699,00 $
- DescriptionFits Flask
- 22877-056ProCulture™ Fittings For Media Handling, with Two 0.6 cm (1/4") Inlets8 L or 15 LCase of 1 - 735,00 $
- Description
- DescriptionFits Flask
- 22877-148ProCulture™ Fittings For Media Handling, with 1.3 cm (1/2") Inlet36 LCase of 1 - 628,00 $
- Description