Corning® Spinner Flasks with Angled Sidearms, Corning
8 L, 120 mm, 45 mm
Supplier: Corning
Supplier #: 4502-8L
Jump to:
Product Details & Documents
Borosilicate glass with side arms, non sterile.
- Baffles enhance aeration and agitation of flask contents
- Impeller design ensures optimal stirring
- Complete with impeller and caps
- Side arm design permit easy access of 25 and 50 ml pipetes
Documents
Spécifications
- Capacité:8 L
- Sidearm Neck O.D.:45 mm
- Center Neck O.D.:120 mm
Accessories
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube89089-9786 LLog in pour la disponibilitéCase of 1 - $676.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long TubesGas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes89090-0026 LLog in pour la disponibilitéCase of 1 - $575.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long TubesGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes89089-98015 LLog in pour la disponibilitéCase of 1 - $676.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long TubesGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes89089-9886 LLog in pour la disponibilitéCase of 1 - $592.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube89089-9966 LLog in pour la disponibilitéCase of 1 - $597.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With 3.2 mm (1/8") and 6.4 (1/4") Inlets and Two Long TubesGas and Media Handling Fittings With 3.2 mm (1/8") and 6.4 (1/4") Inlets and Two Long Tubes89089-9686 LLog in pour la disponibilitéCase of 1 - $626.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube89089-9946 LLog in pour la disponibilitéCase of 1 - $581.00
Informations sur le produit
- Catalog #FitsAvailabilityPrice
- Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)89090-230Corning® Proculture® flasksLog in pour la disponibilitéCase of 1 - $4,070.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Corning® 1 L Proculture® Spinner Flask Gas and Media Handling FittingCorning® 1 L Proculture® Spinner Flask Gas and Media Handling Fitting89090-0061 L angled sidearm spinner flasksLog in pour la disponibilitéCase of 1 - $605.00
Informations sur le produit
- Catalog #FitsAvailabilityPrice
- Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)89090-232Corning® Proculture® flasksLog in pour la disponibilitéCase of 1 - $5,140.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Corning® 3L Proculture® Spinner Flask Gas and Media Handling FittingCorning® 3L Proculture® Spinner Flask Gas and Media Handling Fitting89090-0083 L angled sidearm spinner flasksLog in pour la disponibilitéCase of 1 - $1,130.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long TubesGas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes89089-9981 LLog in pour la disponibilitéCase of 1 - $559.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube89089-9721 LLog in pour la disponibilitéCase of 1 - $553.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4mm (1/4") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 6.4mm (1/4") Inlets, One Short Tube, and One Long Tube89089-97015 LLog in pour la disponibilitéCase of 1 - $597.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube89089-9901 LLog in pour la disponibilitéCase of 1 - $644.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long TubesGas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes89090-0106 LLog in pour la disponibilitéCase of 1 - $630.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube89089-9743 LLog in pour la disponibilitéCase of 1 - $560.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube89089-9923 LLog in pour la disponibilitéCase of 1 - $598.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long TubesGas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes89090-0126 LLog in pour la disponibilitéCase of 1 - $623.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long TubesGas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes89090-0046 LLog in pour la disponibilitéCase of 1 - $592.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long TubesGas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes89089-966500 mlLog in pour la disponibilitéCase of 1 - $612.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long TubeGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube89089-9766 LLog in pour la disponibilitéCase of 1 - $575.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long TubesGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes89089-9843 LLog in pour la disponibilitéCase of 1 - $560.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long TubesGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes89089-9866 LLog in pour la disponibilitéCase of 1 - $575.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long TubesGas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes89089-9821 LLog in pour la disponibilitéCase of 1 - $553.00
Informations sur le produit
- Catalog #Fits FlaskAvailabilityPrice
- Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long TubesGas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes89089-9641 LLog in pour la disponibilitéCase of 1 - $620.00