Corning® Spinner Flasks with Angled Sidearms, Corning
Corning® Spinner Flasks with Angled Sidearms, Corning
8 L, 100 mm, 45 mm
Catalog # 22877-084
Supplier:   Corning
Supplier #:  4500-8L
Corning® Spinner Flasks with Angled Sidearms, Corning
Catalog # 22877-084
Supplier:  Corning
Corning® Spinner Flasks with Angled Sidearms, Corning
Corning® Spinner Flasks with Angled Sidearms, Corning
8 L, 100 mm, 45 mm
Price
$2,490.00
Log in pour la disponibilité
Case of 1
Product Details & Documents
Borosilicate glass with side arms, non sterile.
  • Baffles enhance aeration and agitation of flask contents
  • Impeller design ensures optimal stirring
  • Complete with impeller and caps
  • Side arm design permit easy access of 25 and 50 ml pipetes
Spécifications
  • Capacité:
    8 L
  • Sidearm Neck O.D.:
    45 mm
  • Center Neck O.D.:
    100 mm
Accessories
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    89089-978
    6 L
    Log in pour la disponibilité
    Case of 1 - $676.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    89090-002
    6 L
    Log in pour la disponibilité
    Case of 1 - $575.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    89089-980
    15 L
    Log in pour la disponibilité
    Case of 1 - $676.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    89089-988
    6 L
    Log in pour la disponibilité
    Case of 1 - $592.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    89089-996
    6 L
    Log in pour la disponibilité
    Case of 1 - $597.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With 3.2 mm (1/8") and 6.4 (1/4") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With 3.2 mm (1/8") and 6.4 (1/4") Inlets and Two Long Tubes
    89089-968
    6 L
    Log in pour la disponibilité
    Case of 1 - $626.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    89089-994
    6 L
    Log in pour la disponibilité
    Case of 1 - $581.00
Informations sur le produit
  • Catalog #
    Fits
    Availability
    Price
  • Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)
    89090-230
    Corning® Proculture® flasks
    Log in pour la disponibilité
    Case of 1 - $4,070.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Corning® 1 L Proculture® Spinner Flask Gas and Media Handling Fitting
    Corning® 1 L Proculture® Spinner Flask Gas and Media Handling Fitting
    89090-006
    1 L angled sidearm spinner flasks
    Log in pour la disponibilité
    Case of 1 - $605.00
Informations sur le produit
  • Catalog #
    Fits
    Availability
    Price
  • Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)
    89090-232
    Corning® Proculture® flasks
    Log in pour la disponibilité
    Case of 1 - $5,140.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Corning® 3L Proculture® Spinner Flask Gas and Media Handling Fitting
    Corning® 3L Proculture® Spinner Flask Gas and Media Handling Fitting
    89090-008
    3 L angled sidearm spinner flasks
    Log in pour la disponibilité
    Case of 1 - $1,130.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    89089-998
    1 L
    Log in pour la disponibilité
    Case of 1 - $559.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    89089-972
    1 L
    Log in pour la disponibilité
    Case of 1 - $553.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4mm (1/4") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 6.4mm (1/4") Inlets, One Short Tube, and One Long Tube
    89089-970
    15 L
    Log in pour la disponibilité
    Case of 1 - $597.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    89089-990
    1 L
    Log in pour la disponibilité
    Case of 1 - $644.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes
    Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes
    89090-010
    6 L
    Log in pour la disponibilité
    Case of 1 - $630.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    89089-974
    3 L
    Log in pour la disponibilité
    Case of 1 - $560.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    89089-992
    3 L
    Log in pour la disponibilité
    Case of 1 - $598.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes
    Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes
    89090-012
    6 L
    Log in pour la disponibilité
    Case of 1 - $623.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    89090-004
    6 L
    Log in pour la disponibilité
    Case of 1 - $592.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes
    Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes
    89089-966
    500 ml
    Log in pour la disponibilité
    Case of 1 - $612.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    89089-976
    6 L
    Log in pour la disponibilité
    Case of 1 - $575.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    89089-984
    3 L
    Log in pour la disponibilité
    Case of 1 - $560.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    89089-986
    6 L
    Log in pour la disponibilité
    Case of 1 - $575.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    89089-982
    1 L
    Log in pour la disponibilité
    Case of 1 - $553.00
Informations sur le produit
  • Catalog #
    Fits Flask
    Availability
    Price
  • Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes
    Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes
    89089-964
    1 L
    Log in pour la disponibilité
    Case of 1 - $620.00