Corning® Spinner Flasks with Angled Sidearms, Corning
6 L, 100 mm, 45 mm
: Corning
: 4500-6L
Borosilicate glass with side arms, non sterile.
- Baffles enhance aeration and agitation of flask contents
- Impeller design ensures optimal stirring
- Complete with impeller and caps
- Side arm design permit easy access of 25 and 50 ml pipetes
- Capacité:6 L
- Sidearm Neck O.D.:45 mm
- Center Neck O.D.:100 mm
Accessories
- DescriptionFits Flask
- 89089-978Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube6 LCase of 1 - 676,00 $
- DescriptionFits Flask
- 89090-002Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes6 LCase of 1 - 575,00 $
- DescriptionFits Flask
- 89089-980Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes15 LCase of 1 - 676,00 $
- DescriptionFits Flask
- 89089-988Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes6 LCase of 1 - 592,00 $
- DescriptionFits Flask
- 89089-996Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube6 LCase of 1 - 597,00 $
- DescriptionFits Flask
- 89089-968Gas and Media Handling Fittings With 3.2 mm (1/8") and 6.4 (1/4") Inlets and Two Long Tubes6 LCase of 1 - 626,00 $
- DescriptionFits Flask
- 89089-994Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube6 LCase of 1 - 581,00 $
- DescriptionFits
- 89090-230Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® flasksCase of 1 - 4 070,00 $
- DescriptionFits Flask
- 89090-006Corning® 1 L Proculture® Spinner Flask Gas and Media Handling Fitting1 L angled sidearm spinner flasksCase of 1 - 605,00 $
- DescriptionFits
- 89090-232Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)Corning® Proculture® flasksCase of 1 - 5 140,00 $
- DescriptionFits Flask
- 89090-008Corning® 3L Proculture® Spinner Flask Gas and Media Handling Fitting3 L angled sidearm spinner flasksCase of 1 - 1 130,00 $
- DescriptionFits Flask
- 89089-998Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes1 LCase of 1 - 559,00 $
- DescriptionFits Flask
- 89089-972Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube1 LCase of 1 - 553,00 $
- DescriptionFits Flask
- 89089-970Gas and Media Handling Fittings With Two 6.4mm (1/4") Inlets, One Short Tube, and One Long Tube15 LCase of 1 - 597,00 $
- DescriptionFits Flask
- 89089-990Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube1 LCase of 1 - 644,00 $
- DescriptionFits Flask
- 89090-010Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes6 LCase of 1 - 630,00 $
- DescriptionFits Flask
- 89089-974Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube3 LCase of 1 - 560,00 $
- DescriptionFits Flask
- 89089-992Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube3 LCase of 1 - 598,00 $
- DescriptionFits Flask
- 89090-012Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes6 LCase of 1 - 623,00 $
- DescriptionFits Flask
- 89090-004Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes6 LCase of 1 - 592,00 $
- DescriptionFits Flask
- 89089-966Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes500 mlCase of 1 - 612,00 $
- DescriptionFits Flask
- 89089-976Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube6 LCase of 1 - 575,00 $
- DescriptionFits Flask
- 89089-984Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes3 LCase of 1 - 560,00 $
- DescriptionFits Flask
- 89089-986Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes6 LCase of 1 - 575,00 $
- DescriptionFits Flask
- 89089-982Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes1 LCase of 1 - 553,00 $
- DescriptionFits Flask
- 89089-964Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes1 LCase of 1 - 620,00 $