Corning® Spinner Flasks with Angled Sidearms, Corning
3 L, 100 mm, 45 mm
  22877-080
 :   Corning
 :  4500-3L
Corning® Spinner Flasks with Angled Sidearms, Corning
  22877-080
 :  Corning
Corning® Spinner Flasks with Angled Sidearms, Corning
Corning® Spinner Flasks with Angled Sidearms, Corning
3 L, 100 mm, 45 mm
$1,790.00
Case of 1
Borosilicate glass with side arms, non sterile.
  • Baffles enhance aeration and agitation of flask contents
  • Impeller design ensures optimal stirring
  • Complete with impeller and caps
  • Side arm design permit easy access of 25 and 50 ml pipetes
  • Capacité:
    3 L
  • Sidearm Neck O.D.:
    45 mm
  • Center Neck O.D.:
    100 mm
Accessories
  •  
    Description
    Fits Flask
  • 89089-978
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    6 L
    Case of 1 - $676.00
  •  
    Description
    Fits Flask
  • 89090-002
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    6 L
    Case of 1 - $575.00
  •  
    Description
    Fits Flask
  • 89089-980
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    15 L
    Case of 1 - $676.00
  •  
    Description
    Fits Flask
  • 89089-988
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    6 L
    Case of 1 - $592.00
  •  
    Description
    Fits Flask
  • 89089-996
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    6 L
    Case of 1 - $597.00
  •  
    Description
    Fits Flask
  • 89089-968
    Gas and Media Handling Fittings With 3.2 mm (1/8") and 6.4 (1/4") Inlets and Two Long Tubes
    6 L
    Case of 1 - $626.00
  •  
    Description
    Fits Flask
  • 89089-994
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    6 L
    Case of 1 - $581.00
  •  
    Description
    Fits
  • 89090-230
    Corning® Proculture® Spinner Direct Top Drive Motor, 120 VAC 60 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® flasks
    Case of 1 - $4,070.00
  •  
    Description
    Fits Flask
  • 89090-006
    Corning® 1 L Proculture® Spinner Flask Gas and Media Handling Fitting
    1 L angled sidearm spinner flasks
    Case of 1 - $605.00
  •  
    Description
    Fits
  • 89090-232
    Corning® Proculture® Spinner Direct Top Drive Motor, 230 VAC 50 Hz, 4.1 Kg (9 lbs.)
    Corning® Proculture® flasks
    Case of 1 - $5,140.00
  •  
    Description
    Fits Flask
  • 89090-008
    Corning® 3L Proculture® Spinner Flask Gas and Media Handling Fitting
    3 L angled sidearm spinner flasks
    Case of 1 - $1,130.00
  •  
    Description
    Fits Flask
  • 89089-998
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    1 L
    Case of 1 - $559.00
  •  
    Description
    Fits Flask
  • 89089-972
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    1 L
    Case of 1 - $553.00
  •  
    Description
    Fits Flask
  • 89089-970
    Gas and Media Handling Fittings With Two 6.4mm (1/4") Inlets, One Short Tube, and One Long Tube
    15 L
    Case of 1 - $597.00
  •  
    Description
    Fits Flask
  • 89089-990
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    1 L
    Case of 1 - $644.00
  •  
    Description
    Fits Flask
  • 89090-010
    Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes
    6 L
    Case of 1 - $630.00
  •  
    Description
    Fits Flask
  • 89089-974
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    3 L
    Case of 1 - $560.00
  •  
    Description
    Fits Flask
  • 89089-992
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets, One Short Tube, and One Long Tube
    3 L
    Case of 1 - $598.00
  •  
    Description
    Fits Flask
  • 89090-012
    Gas and Media Handling Fittings With Three 3.2 mm (1/8") Inlets, One Short Tube, and Two Long Tubes
    6 L
    Case of 1 - $623.00
  •  
    Description
    Fits Flask
  • 89090-004
    Gas and Media Handling Fittings With Two 3.2 mm (1/8") Inlets and Two Long Tubes
    6 L
    Case of 1 - $592.00
  •  
    Description
    Fits Flask
  • 89089-966
    Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes
    500 ml
    Case of 1 - $612.00
  •  
    Description
    Fits Flask
  • 89089-976
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets, One Short Tube, and One Long Tube
    6 L
    Case of 1 - $575.00
  •  
    Description
    Fits Flask
  • 89089-984
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    3 L
    Case of 1 - $560.00
  •  
    Description
    Fits Flask
  • 89089-986
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    6 L
    Case of 1 - $575.00
  •  
    Description
    Fits Flask
  • 89089-982
    Gas and Media Handling Fittings With Two 6.4 mm (1/4") Inlets and Two Long Tubes
    1 L
    Case of 1 - $553.00
  •  
    Description
    Fits Flask
  • 89089-964
    Gas and Media Handling Fittings With One 3.2 mm (1/8") Inlet, One 6.4 mm (1/4") Inlet, and Two Long Tubes
    1 L
    Case of 1 - $620.00