Iris Scissors, Delicate, Integra™ Miltex® | Avantor
Iris Scissors, Delicate, Integra™ Miltex®
Curved, 89 mm (3½"), 20 mm (¾")
Catalog # 21909-246
Supplier:   Miltex
Fournisseur #:  18-1398
Iris Scissors, Delicate, Integra™ Miltex®
Catalog # 21909-246
Supplier:  Miltex
Iris Scissors, Delicate, Integra™ Miltex®
Iris Scissors, Delicate, Integra™ Miltex®
Curved, 89 mm (3½"), 20 mm (¾")
Price
$343.69
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Delicate blades with sharp/sharp tips
Stainless steel.
Spécifications
  • Désignation:
    Curved
  • Longueur:
    89 mm (3½")
  • Blade Length:
    20 mm (¾")