VWR® Sharps Container Systems, Red | Avantor
VWR® Sharps Container Systems, Red
5.75 gal., Non-Stackable, 30.5W×22.9D×45.7H cm (12W×9D×18"H), Red
Catalog # 19001-030
Supplier:   VWR International
VWR® Sharps Container Systems, Red
Catalog # 19001-030
Supplier:  VWR International
VWR® Sharps Container Systems, Red
VWR® Sharps Container Systems, Red
5.75 gal., Non-Stackable, 30.5W×22.9D×45.7H cm (12W×9D×18"H), Red
Price
$372.61
Log in pour la disponibilité
Each Case of 4
Product Details & Documents
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps.
  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The stackable sharps containers are puncture-resistant and both autoclaves-safe and incinerator-safe. The tortuous path systems restrict hand entry. The extended neck sharps containers feature a funneled vertical drop and extended handles.
Spécifications
  • Capacité:
    5.75 gal.
  • Color:
    Red
  • Dimensions:
    30.5W×22.9D×45.7H cm (12W×9D×18"H)
  • Style:
    Non-Stackable